1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
Thermo Scientific Chemicals
1-Methylnaphthalene, 96%
CAS: 90-12-0 | C11H10 | 142.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA127165000
Price (MYR)
721.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
721.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Methylnaphthalene
Name Note97%
CAS90-12-0
CAS Min %97+%
Health Hazard 1GHS Signal Word: Warning