2-(Methylthio)aniline, 97%
2-(Methylthio)aniline, 97%
2-(Methylthio)aniline, 97%
2-(Methylthio)aniline, 97%
Thermo Scientific Chemicals
2-(Methylthio)aniline, 97%
CAS: 2987-53-3 | C7H9NS | 139.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA127110250
Price (MYR)
1,027.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,027.00
EA
Add to cart
Specifications
Chemical Name or Material2-(Methylthio)aniline
CAS2987-53-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0