Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Methyl 4-hydroxybenzoate, 99%
Thermo Scientific Chemicals
Methyl 4-hydroxybenzoate, 99%
CAS: 99-76-3 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA126961000100 gPlastic bottle
Catalog number FSA126961000
Price (MYR)
246.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
246.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl 4-hydroxybenzoate
Name Note99%
CAS99-76-3
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.