4,4'-Methylenebis(N,N-dimethylaniline), 98%
4,4'-Methylenebis(N,N-dimethylaniline), 98%
4,4'-Methylenebis(N,N-dimethylaniline), 98%
4,4'-Methylenebis(N,N-dimethylaniline), 98%
Thermo Scientific Chemicals
4,4'-Methylenebis(N,N-dimethylaniline), 98%
CAS: 101-61-1 | C17H22N2 | 254.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA126745000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4, 4'-Methylenebis(N, N-dimethylaniline)
Name Note99+%
CAS101-61-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger