Specifications
Chemical Name or MaterialMethylcyclohexane
Name Note99%
CAS108-87-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
View more