Methyl 4-chlorobutyrate, 98+%
Methyl 4-chlorobutyrate, 98+%
Methyl 4-chlorobutyrate, 98+%
Methyl 4-chlorobutyrate, 98+%
Thermo Scientific Chemicals
Methyl 4-chlorobutyrate, 98+%
CAS: 3153-37-5 | C5H9ClO2 | 136.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA126521000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl 4-chlorobutyrate
Name Note98+%
CAS3153-37-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.