2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%
Thermo Scientific Chemicals
2-Methyl-2-butene, 99+%
2-Methyl-2-butene, 99+%, C5H10, CAS Number-513-35-9, 1,1,2-trimethylethylene, 2-butene, 2-methyl, 2-methyl-2-butene, 2-methylbutene-2, 3-methyl-2-butene, amylene, beta-isoamylene, ethylene, trimethyl, n-amylene, trimethylethylene, 250mL, 99% min. (GC), 01, 211, 35.0 deg.C to 38.0 deg.C, 100.0, 99.0 | CAS: 513-35-9 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA126490250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-2-butene
Name Note99+%
CAS513-35-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger