9-Methylanthracene, 99%
9-Methylanthracene, 99%
9-Methylanthracene, 99%
9-Methylanthracene, 99%
Thermo Scientific Chemicals
9-Methylanthracene, 99%
CAS: 779-02-2 | C15H12 | 192.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA126300050
Price (MYR)
678.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
678.00
BOT
Add to cart
Specifications
Chemical Name or Material9-Methylanthracene
CAS779-02-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici