Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA126195000
Price (MYR)
361.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
361.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl acrylate
Name NoteStabilized
CAS96-33-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic if inhaled