3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
3'-Methylacetophenone, 97%
CAS: 585-74-0 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA126170250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3′-Methylacetophenone
CAS585-74-0
CAS Min %96.0
Health Hazard 1Warning
CAS Max %100.0