Check Order Status
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number
FSA126132500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
5-Methoxytryptamine hydrochloride
Name Note
97%
CAS
66-83-1
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning