4-Methoxyphenylacetic acid, 99%
4-Methoxyphenylacetic acid, 99%
4-Methoxyphenylacetic acid, 99%
4-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals
4-Methoxyphenylacetic acid, 99%
CAS: 104-01-8 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number FSA126022500
Price (MYR)
562.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
562.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenylacetic acid
Name Note99%
CAS104-01-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning