3-Methoxyphenylacetic acid, 99.5%
3-Methoxyphenylacetic acid, 99.5%
3-Methoxyphenylacetic acid, 99.5%
3-Methoxyphenylacetic acid, 99.5%
Thermo Scientific Chemicals
3-Methoxyphenylacetic acid, 99.5%
CAS: 1798-09-0 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA126010250
Price (MYR)
668.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
668.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Methoxyphenylacetic acid
CAS1798-09-0
CAS Min %99.4
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0