4-Methoxyphenol, 99%
4-Methoxyphenol, 99%
4-Methoxyphenol, 99%
4-Methoxyphenol, 99%
Thermo Scientific Chemicals
4-Methoxyphenol, 99%
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
5 kg
Catalog number FSA126005000
Price (MYR)
570.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
570.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenol
Name NoteStabilized
CAS150-76-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning