2-Mercaptothiazoline, 98%
2-Mercaptothiazoline, 98%
2-Mercaptothiazoline, 98%
2-Mercaptothiazoline, 98%
Thermo Scientific Chemicals
2-Mercaptothiazoline, 98%
CAS: 96-53-7 | C3H5NS2 | 119.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA125560050
Price (MYR)
195.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
195.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptothiazoline
Name Note98%
CAS96-53-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning