2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
2-Mercaptoethanol, 99%, pure
Thermo Scientific Chemicals
2-Mercaptoethanol, 99%, pure
CAS: 60-24-2 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA125472500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Mercaptoethanol
CAS60-24-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause damage to organs through prolon
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat