L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
Thermo Scientific Chemicals
L(-)-Menthol, 99.5%
CAS: 2216-51-5 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA125405000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(-)-Menthol
Name Note99.7%
CAS2216-51-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement