Malonyl dichloride, 97%
Malonyl dichloride, 97%
Malonyl dichloride, 97%
Malonyl dichloride, 97%
Thermo Scientific Chemicals
Malonyl dichloride, 97%
CAS: 1663-67-8 | C3H2Cl2O2 | 140.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA125280250
Price (MYR)
974.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
974.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMalonyl dichloride
CAS1663-67-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre