Linalool, 97%
Linalool, 97%
Linalool, 97%
Thermo Scientific Chemicals
Linalool, 97%
CAS: 78-70-6 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA125155000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLinalool
Name Note97%
CAS78-70-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement