L-Leucine, 99%
L-Leucine, 99%
L-Leucine, 99%
L-Leucine, 99%
L-Leucine, CAS # 61-90-5, is a branched-chain aliphatic essential amino acid in humans that is important for protein synthesis and many metabolic functions. | CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA125121000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Leucine
Melting Point286.0°C to 288.0°C
CAS61-90-5
Molecular FormulaC6H13NO2
MDL NumberMFCD00002617
View more
General Description
  • L-Leucine is an essential amino acid that has an important role in the growth and repair of muscle and bone tissue
  • It functions to preserve muscle glycogen and helps to prevent the breakdown of muscle proteins after trauma or severe stress
  • It is also involved in hemoglobin formation and metabolic functions. This amino acid cannot be synthesized to meet the body's needs from simpler molecules, and therefore must be provided in the diet.
  • L-Leucine stimulates protein synthesis by the initiation of mRNA translation
Applications
  • L-Leucine is used in the biosynthesis of proteins
  • In vivo studies show that L-Leucine can be used when gene defects lead to an organism's inability to metabolize the amino acid phenylalanine
  • This compound also contributes to the regulation of blood-sugar levels, growth hormone production, and wound healing
  • In research experiments this compound has been used as an activator of the mammalian target of rapamycin (mTOR)
RUO – Research Use Only