L(+)-Lactic acid, 20 wt.% solution in water
L(+)-Lactic acid, 20 wt.% solution in water
L(+)-Lactic acid, 20 wt.% solution in water
L(+)-Lactic acid, 20 wt.% solution in water
Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA125070025
Price (MYR)
722.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
722.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Lactic acid
CAS79-33-4
Health Hazard 1Danger
ColorColorless
Health Hazard 2GHS P Statement