Citric acid monohydrate, 99.5%, for analysis
Citric acid monohydrate, 99.5%, for analysis
Citric acid monohydrate, 99.5%, for analysis
Citric acid monohydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis
CAS: 5949-29-1 | C6H10O8 | 210.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA124912500
Price (MYR)
237.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
237.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitric acid monohydrate
CAS5949-29-1
Melting Point135.0°C to 152.0°C
Health Hazard 1Warning
ColorWhite
View more
Biological buffer for analysis
RUO – Research Use Only