Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Thermo Scientific Chemicals
Hydrochloric acid, for analysis, fuming, 37% solution in water
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA124630010
Price (MYR)
248.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
248.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Melting Point-35.0°C
Boiling Point57.0°C
Health Hazard 1Danger
View more
For analysis
RUO – Research Use Only