Fluoxymesterone, 97.0-102.0%
Fluoxymesterone, 97.0-102.0%
Fluoxymesterone, 97.0-102.0%
Fluoxymesterone, 97.0-102.0%
Thermo Scientific Chemicals
Fluoxymesterone, 97.0-102.0%
Fluoxymesterone, CAS # 76-43-7, is a potent, orally administered, synthetic steroid with androgenic activity. It is used in the treatment of male hypogonadism. | CAS: 76-43-7 | C20H29FO3 | 336.447 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA124590050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFluoxymesterone
CAS76-43-7
Melting Point240.0°C
PackagingGlass bottle
Molecular FormulaC20H29FO3
View more
General description
  • Fluoxymesterone is a white, odorless, crystalline powder, soluble in pyridine
  • It is an orally-administered synthetic steroid that is about 5-10 times more potent than male steroid, testosterone
  • It is metabolized in the liver, and less than 5% of this drug is excreted from the body unchanged
  • Adverse effects of fluoxymesterone include sodium and water retention leading to edema, acne, cardiovascular toxicity, and hepatotoxicity
Applications
  • Fluoxymesterone has been found to competitively inhibit prolactin or estrogen receptors in tumorigenic breast cells, and is therefore administered in the treatment of breast cancer
  • In males, this compound is used as a replacement in conditions associated with symptoms of deficiency or absence of endogenous testosterone
RUO – Research Use Only