beta-Nicotinamide adenine dinucleotide hydrate, 98+%
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide hydrate, 98+%
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA124532500
Price (MYR)
367.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
367.00
BOT
Add to cart
Specifications
Chemical Name or Materialβ-Nicotinamide adenine dinucleotide hydrate
CAS53-84-9
CAS Min %98.0
CAS Max %100.0
CAS53-84-9