Oxalic acid, 0.1 N standard solution
Oxalic acid, 0.1 N standard solution
Oxalic acid, 0.1 N standard solution
Oxalic acid, 0.1 N standard solution
Thermo Scientific Chemicals
Oxalic acid, 0.1 N standard solution
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number FSA124280025
Price (MYR)
458.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
458.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxalic acid
CAS144-62-7
ColorColorless
Molecular FormulaC2H2O4
Linear FormulaHO2CCO2H
View more
Volumetric standard solution
RUO – Research Use Only