Iodine, 0.1N (0.05M) standard solution
Iodine, 0.1N (0.05M) standard solution
Iodine, 0.1N (0.05M) standard solution
Iodine, 0.1N (0.05M) standard solution
Thermo Scientific Chemicals
Iodine, 0.1N (0.05M) standard solution
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA124220025
Price (MYR)
1,172.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,172.00
EA
Add to cart
Specifications
Chemical Name or MaterialIodine
Name Note0.1N Standard Solution
CAS7553-56-2
ColorBrown
Health Hazard 2GHS H Statement:
Harmful to aquatic life with long lasting effects.
View more
Volumetric standard solution
RUO – Research Use Only