Sodium hydroxide, 0.1 N standard solution
Sodium hydroxide, 0.1 N standard solution
Sodium hydroxide, 0.1 N standard solution
Sodium hydroxide, 0.1 N standard solution
Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution
Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL | CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Plastic Bottle
Catalog number FSA124190025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2, 7732-18-5
Specifications
Chemical Name or MaterialSodium hydroxide
ColorWhite to Beige
Name Note0.1 N Standard Solution
CAS1310-73-2
CAS7732-18-5
View more
Volumetric standard solution
RUO – Research Use Only