Acetic acid, 99.5%, pure
Acetic acid, 99.5%, pure
Acetic acid, 99.5%, pure
Acetic acid, 99.5%, pure
Thermo Scientific Chemicals
Acetic acid, 99.5%, pure
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA124040025
Price (MYR)
669.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
669.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic acid
CAS64-19-7
Melting Point16.0°C to 16.5°C
Boiling Point117.0°C to 118.0°C
CAS Min %99.4