Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
Thermo Scientific Chemicals
Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA124020025
Price (MYR)
666.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
666.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMineral oil
CAS8042-47-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
View more
For IR spectroscopy
RUO – Research Use Only