Diethyl ether, 99+%, pure, stabilized with BHT
Diethyl ether, 99+%, pure, stabilized with BHT
Diethyl ether, 99+%, pure, stabilized with BHT
Diethyl ether, 99+%, pure, stabilized with BHT
Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA123990025
Price (MYR)
580.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
580.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethyl ether
Melting Point-116.0°C
CAS60-29-7
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.