4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
Thermo Scientific Chemicals
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
CAS: 4940-39-0 | C10H6O4 | 190.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA123880050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Oxo-4H-1-benzopyran-2-carboxylic acid
Name Note97%
CAS4940-39-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning