Oxytetracycline Dihydrate 95%
Oxytetracycline Dihydrate 95%
Oxytetracycline Dihydrate 95%
Oxytetracycline Dihydrate 95%
Thermo Scientific Chemicals
Oxytetracycline Dihydrate 95%
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass Bottle
Catalog number FSA123841000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOxytetracycline dihydrate
TypeOxytetracycline Dihydrate
Name Note95%
Physical FormCrystalline Powder
CAS6153-64-6