8-Aminoquinoline, 98+%
8-Aminoquinoline, 98+%
8-Aminoquinoline, 98+%
8-Aminoquinoline, 98+%
Thermo Scientific Chemicals
8-Aminoquinoline, 98+%
CAS: 578-66-5 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA123710100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material8-Aminoquinoline
CAS578-66-5
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0