Hesperidin, 90+%
Hesperidin, 90+%
Hesperidin, 90+%
Thermo Scientific Chemicals
Hesperidin, 90+%
CAS: 520-26-3 | C28H34O15 | 610.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA123462500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHesperidin
Name Note97%
CAS520-26-3
Health Hazard 1HYGROSCOPIC
Melting Point258.0°C to 262.0°C