Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
Thermo Scientific Chemicals
Calcium chloride dihydrate, extra pure, flakes for drying purposes, 77-80% CaCl2
CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
Packaging:
Plastic drum
Catalog number FSA123350100
Price (MYR)
1,505.00
EA
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,505.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride dihydrate
Melting Point175.0°C
Name NoteFor drying pursposes
CAS10035-04-8
ColorGray-White to White
View more
Adsorbent
RUO – Research Use Only