Ammonium chloride, 99+%, pure
Ammonium chloride, 99+%, pure
Ammonium chloride, 99+%, pure
Ammonium chloride, 99+%, pure
Thermo Scientific Chemicals
Ammonium chloride, 99+%, pure
Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure | CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA123340250
Price (MYR)
210.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
210.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340.0°C
CAS12125-02-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning