Isoprene, 98%, stabilized
Isoprene, 98%, stabilized
Isoprene, 98%, stabilized
Isoprene, 98%, stabilized
Thermo Scientific Chemicals
Isoprene, 98%, stabilized
CAS: 78-79-5 | C5H8 | 68.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA122675000
Price (MYR)
581.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
581.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsoprene
CAS78-79-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
CAS98-29-3