Isophthalic acid, 99%
Isophthalic acid, 99%
Isophthalic acid, 99%
Isophthalic acid, 99%
Thermo Scientific Chemicals
Isophthalic acid, 99%
CAS: 121-91-5 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2 kg
Packaging:
View all
Plastic bottle
Catalog number FSA122655000
Price (MYR)
312.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
312.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsophthalic acid
CAS121-91-5
Melting Point341.0°C to 343.0°C
ColorWhite
EINECS Number204-506-4