Isophorone, 97%
Isophorone, 97%
Isophorone, 97%
Thermo Scientific Chemicals
Isophorone, 97%
CAS: 78-59-1 | C9H14O | 138.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA122645000
Price (MYR)
270.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
270.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsophorone
Name Note98%
CAS78-59-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Suspected of causing cancer.