Isonicotinic acid, 99%
Isonicotinic acid, 99%
Isonicotinic acid, 99%
Isonicotinic acid, 99%
Thermo Scientific Chemicals
Isonicotinic acid, 99%
CAS: 55-22-1 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA122595000
Price (MYR)
633.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
633.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsonicotinic acid
CAS55-22-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
View more