Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Thermo Scientific Chemicals
Isobutyric Acid, 99+%
CAS: 79-31-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA122525000
Price (MYR)
448.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
448.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsobutyric acid
Name Note99+%
CAS79-31-2
Melting Point-47.0°C
Boiling Point153.0°C to 154.0°C
View more
Acylation reagent
RUO – Research Use Only