Isobutyraldehyde, 99+%
Isobutyraldehyde, 99+%
Isobutyraldehyde, 99+%
Isobutyraldehyde, 99+%
Thermo Scientific Chemicals
Isobutyraldehyde, 99+%
CAS: 78-84-2 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA122510010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsobutyraldehyde
CAS78-84-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond conta