Iodonitrotetrazolium chloride, 98%
Iodonitrotetrazolium chloride, 98%
Iodonitrotetrazolium chloride, 98%
Iodonitrotetrazolium chloride, 98%
Thermo Scientific Chemicals
Iodonitrotetrazolium chloride, 98%
CAS: 146-68-9 | C19H13ClIN5O2 | 505.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA122400250
Price (MYR)
9,843.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
9,843.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodonitrotetrazolium chloride
CAS146-68-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Health Hazard 3GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a