1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
Thermo Scientific Chemicals
1-Iodo-4-nitrobenzene, 99%
CAS: 636-98-6 | C6H4INO2 | 249.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA122380250
Price (MYR)
806.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
806.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Iodo-4-nitrobenzene
Name Note99%
CAS636-98-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning