Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Thermo Scientific Chemicals
Iodomethane, 99%, stabilized
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA122375000
Price (MYR)
1,129.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,129.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodomethane
CAS74-88-4
Health Hazard 1Signal Word: WARNING!
CAS7440-50-8
Health Hazard 3Emergency Overview
Limited evidence of a carcinogenic effect. Toxic by inhalation and if swallowed. Harmful in contact with skin. Irritating to eyes, respiratory system and skin.
NFPA
View more
Alkylation reagent
RUO – Research Use Only