2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
Thermo Scientific Chemicals
2-Iodobenzoic acid, 98%
CAS: 88-67-5 | C7H5IO2 | 248.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA122341000
Price (MYR)
541.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
541.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Iodobenzoic acid
CAS88-67-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for