4-Iodoanisole, 98%
4-Iodoanisole, 98%
4-Iodoanisole, 98%
Thermo Scientific Chemicals
4-Iodoanisole, 98%
CAS: 696-62-8 | C7H7IO | 234.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA122321000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodoanisole
CAS696-62-8
Melting Point46.0°C to 51.0°C
ColorBrown to White
Recommended StorageProduct may darken on storage