Iodoacetic acid, 97%
Iodoacetic acid, 97%
Iodoacetic acid, 97%
Iodoacetic acid, 97%
Thermo Scientific Chemicals
Iodoacetic acid, 97%
CAS: 64-69-7 | C2H3IO2 | 185.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA122280250
Price (MYR)
541.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
541.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodoacetic acid
Name Note97%
CAS64-69-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement