Iodoacetamide, 98%
Iodoacetamide, 98%
Iodoacetamide, 98%
Thermo Scientific Chemicals
Iodoacetamide, 98%
CAS: 144-48-9 | C2H4INO | 184.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA122271000
Price (MYR)
2,902.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,902.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodoacetamide
CAS144-48-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee